Merck
  • GAS1 is required for NOTCH-dependent facilitation of SHH signaling in the ventral forebrain neuroepithelium.
GAS1 is required for NOTCH-dependent facilitation of SHH signaling in the ventral forebrain neuroepithelium.
Development (Cambridge, England) (2021-10-27)
Maike Marczenke, Daniele Yumi Sunaga-Franze, Oliver Popp, Irene W Althaus, Sascha Sauer, Philipp Mertins, Annabel Christ, Benjamin L Allen, Thomas E Willnow
ABSTRACT
Growth arrest-specific 1 (GAS1) acts as a co-receptor to patched 1, promoting sonic hedgehog (SHH) signaling in the developing nervous system. GAS1 mutations in humans and animal models result in forebrain and craniofacial malformations, defects ascribed to a function for GAS1 in SHH signaling during early neurulation. Here, we confirm loss of SHH activity in the forebrain neuroepithelium in GAS1-deficient mice and in induced pluripotent stem cell-derived cell models of human neuroepithelial differentiation. However, our studies document that this defect can be attributed, at least in part, to a novel role for GAS1 in facilitating NOTCH signaling, which is essential to sustain a persistent SHH activity domain in the forebrain neuroepithelium. GAS1 directly binds NOTCH1, enhancing ligand-induced processing of the NOTCH1 intracellular domain, which drives NOTCH pathway activity in the developing forebrain. Our findings identify a unique role for GAS1 in integrating NOTCH and SHH signal reception in neuroepithelial cells, and they suggest that loss of GAS1-dependent NOTCH1 activation contributes to forebrain malformations in individuals carrying GAS1 mutations.
Sigma-Aldrich
SAG dihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®