Skip to Content
Merck
All Photos(1)

Key Documents

CP06

Sigma-Aldrich

Anti-α-Tubulin Mouse mAb (DM1A)

liquid, clone DM1A, Calbiochem®

Synonym(s):

α-Tubulin Detection Antibody, Anti-Tubulin DM1A, Mouse Anti-α-Tubulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

DM1A, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, mouse, gerbil, chicken, rat

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TUBA1A(7846)

General description

Purified mouse monoclonal antibody generated by immunizing Balb/c57 mice with the specified immunogen and fusing splenocytes with NS-1 mouse myeloma cells. Recognizes the ~60 kDa tubulin protein.
Recognizes the ~60 kDa tubulin protein found in all eukaryotic cells. May be used as a positive control.
This Anti-α-Tubulin Mouse mAb (DM1A) is validated for use in Immunoblotting, Immunofluorescence for the detection of α-Tubulin.

Immunogen

Chicken
native chick brain microtubules

Application

Immunoblotting (20-100 ng/ml, colorimetric or chemiluminescence; see application references)

Immunofluorescence (5 µg/ml)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 50 mM sodium phosphate buffer, 0.2% gelatin.

Analysis Note

Negative Control
trp E (Ab-1)
Positive Control
Any cell line

Other Notes

Blose, S.H., et al. 1984. J. Cell. Biol.98, 847.
Lin, J.J.C., et al. 1984. In: Monoclonal antibodies and functional cell lines. Kennett, R.H., Bechtol, K.B. and McKearn, T.J., eds. Plenum Publishing Corp.
Lin, J.J.C. 1982. Cold Spring Harbor Symp. on Quant. Biol.46, 769.
Goldman, R.D., et al. 1979. Ann. Rev. Physiol.41, 701.
Goldman, R., Pollard, T. and Rosenbaum, J. (eds.) 1976. Cold Spring Harbor Conference on Cell Proliferation, Books A-C, Cold Spring Harbor, New York.
Tubulin is found in all cells and is used as a positive control for immunofluorescence or immunoblotting. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Kremp et al.
Frontiers in cell and developmental biology, 8, 251-251 (2020-05-21)
Proteins destined for transport to specific organelles usually contain targeting information, which are embedded in their sequence. Many enzymes are required in more than one cellular compartment and different molecular mechanisms are used to achieve dual localization. Here we report
Paulina Podkalicka et al.
Biomolecules, 10(1) (2020-01-23)
Elevated expression of heme oxygenase-1 (HO-1, encoded by HMOX1) is observed in various types of tumors. Hence, it is suggested that HO-1 may serve as a potential target in anticancer therapies. A novel approach to inhibit HO-1 is related to
K Ashley Hickman et al.
Cytometry. Part A : the journal of the International Society for Analytical Cytology, 97(4), 363-377 (2019-11-28)
Short half-life proteins regulate many essential processes, including cell cycle, transcription, and apoptosis. However, few well-characterized protein-turnover pathways have been identified because traditional methods to measure protein half-life are time and labor intensive. To overcome this barrier, we developed a
B A Ibrahim et al.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme, 47(8), 611-621 (2014-09-18)
Short-term abstinence from food intake, planned or unplanned, is unavoidable in modern life, but negatively correlated with appetite control and obesity. This study investigated the role of estradiol in feeding and body weight (BW) reactions to short-span cessation of feeding.
L Francisco Lorenzo-Martín et al.
Oncogene, 38(2), 209-227 (2018-08-09)
The bidirectional regulation of epithelial-mesenchymal transitions (EMT) is key in tumorigenesis. Rho GTPases regulate this process via canonical pathways that impinge on the stability of cell-to-cell contacts, cytoskeletal dynamics, and cell invasiveness. Here, we report that the Rho GTPase activators

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service