Merck
  • 2-Deoxy-D-glucose impedes T cell-induced apoptosis of keratinocytes in oral lichen planus.
2-Deoxy-D-glucose impedes T cell-induced apoptosis of keratinocytes in oral lichen planus.
Journal of cellular and molecular medicine (2021-10-22)
Fang Wang, Jing Zhang, Gang Zhou
ABSTRACT
Oral lichen planus (OLP) is a T cell-mediated immunoinflammatory disease. Glycolysis plays an essential role in T-cell immune responses. Blocking glycolytic pathway in activated T cells represents a therapeutic strategy for restraint of immunologic process in autoimmune disorders. 2-Deoxy-D-glucose (2-DG) has been widely used to probe into glycolysis in immune cells. This study was aimed to explore the role of glycolysis inhibition by 2-DG on regulating immune responses of OLP-derived T cells. We observed that lactic dehydrogenase A (LDHA) expression was elevated in OLP lesions and local T cells. 2-DG inhibited the expression of LDHA, p-mTOR, Hif1α and PLD2 in T cells; meanwhile, it decreased proliferation and increased apoptosis of T cells. T cells treated by 2-DG showed lower LDHA expression and elevated apoptosis, resulting in a reduced apoptotic population of keratinocytes that were co-cultured with them, which was related to the decreased levels of IFN-γ in co-culture system. Rapamycin enhanced the effects of 2-DG on immune responses between T cells and keratinocytes. Thus, these findings indicated that OLP-derived T cells might be highly dependent upon high glycolysis for proliferation, and 2-DG treatment combined with rapamycin might be an option to alleviate T-cell responses, contributing to reducing apoptosis of keratinocytes.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, γ-irradiated, BioXtra, suitable for cell culture