Merck
  • NDUFS3 depletion permits complex I maturation and reveals TMEM126A/OPA7 as an assembly factor binding the ND4-module intermediate.
NDUFS3 depletion permits complex I maturation and reveals TMEM126A/OPA7 as an assembly factor binding the ND4-module intermediate.
Cell reports (2021-04-22)
Luigi D'Angelo, Elisa Astro, Monica De Luise, Ivana Kurelac, Nikkitha Umesh-Ganesh, Shujing Ding, Ian M Fearnley, Giuseppe Gasparre, Massimo Zeviani, Anna Maria Porcelli, Erika Fernandez-Vizarra, Luisa Iommarini
ABSTRACT
Complex I (CI) is the largest enzyme of the mitochondrial respiratory chain, and its defects are the main cause of mitochondrial disease. To understand the mechanisms regulating the extremely intricate biogenesis of this fundamental bioenergetic machine, we analyze the structural and functional consequences of the ablation of NDUFS3, a non-catalytic core subunit. We show that, in diverse mammalian cell types, a small amount of functional CI can still be detected in the complete absence of NDUFS3. In addition, we determine the dynamics of CI disassembly when the amount of NDUFS3 is gradually decreased. The process of degradation of the complex occurs in a hierarchical and modular fashion in which the ND4 module remains stable and bound to TMEM126A. We, thus, uncover the function of TMEM126A, the product of a disease gene causing recessive optic atrophy as a factor necessary for the correct assembly and function of CI.
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
G 418 disulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
GenElute Mammalian Genomic DNA Miniprep Kits, sufficient for 350 purifications
Millipore
Digitonin, High Purity, CAS 11024-24-1 is a non-ionic detergent commonly used to solubilize membrane-bound proteins.
Sigma-Aldrich
Lysis solution for blood, sufficient for 100 reactions, Molecular Biology
Sigma-Aldrich
L-Lysine-13C6,15N2 hydrochloride, 99 atom % 13C, 99 atom % 15N, 95% (CP)
Sigma-Aldrich
L-Arginine-13C6,15N4 hydrochloride, 99 atom % 13C, 99 atom % 15N, 95% (CP)
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) disodium salt hydrate, vial of ~1 mg ATP
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) assay mix, lyophilized powder
Sigma-Aldrich
Neutralization Solution for Blood, sufficient for 100 reactions, Molecular Biology, sufficient for 1000 reactions
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture