Merck
  • Comparing micellar, hemolytic, and antibacterial properties of di- and tricarboxyl dendritic amphiphiles.
Comparing micellar, hemolytic, and antibacterial properties of di- and tricarboxyl dendritic amphiphiles.
Bioorganic & medicinal chemistry (2011-04-16)
Bhadreshkumar B Maisuria, Marcelo L Actis, Shauntrece N Hardrict, Joseph O Falkinham, Michael F Cole, Ronald L Cihlar, Stephen M Peters, Richard V Macri, Eko W Sugandhi, André A Williams, Michael A Poppe, Alan R Esker, Richard D Gandour
ABSTRACT
Homologous dicarboxyl dendritic amphiphiles-RCONHC(CH(3))(CH(2)CH(2)COOH)(2), 4(n); and ROCONHC(CH(3))(CH(2)CH(2)COOH)(2), 5(n), where R=n-C(n)H(2)(n)(+1) and n=13-22 carbon atoms-were synthesized. Critical micelle concentrations (CMCs) in aqueous triethanolamine solutions and at pH 7.4 were measured along with hemolytic activity (effective concentrations, EC(10)) in phosphate-buffered saline (PBS). LogCMC showed a linear dependence on chain length (n); the longest chain in each series had the lowest CMC-in triethanolamine: 4(21), 180μM and 5(22), 74μM and at pH 7.4: 4(21), 78μM and 5(22), 33μM. These two series, 4(n) and 5(n), and three series of homologous tricarboxyl dendritic amphiphiles-RCONHC(CH(2)CH(2)COOH)(3), 1(n); ROCONHC(CH(2)CH(2)COOH)(3), 2(n); RNHCONHC(CH(2)CH(2)COOH)(3), 3(n), where R=n-C(n)H(2)(n)(+1) and n=13-22 carbon atoms-were tested for growth inhibition of Staphylococcus aureus strain ATCC 6358 and methicillin-resistant S. aureus (MRSA) strain ATCC 43330 by microdilution in 0.1-strength brain heart infusion broth (BHIB). Amphiphiles 4(19), 4(21), 5(18), and 5(20) showed the strongest antibacterial activity (2.2-3.4μg/mL) against S. aureus (vancomycin, MIC=0.25μg/mL). These four plus 1(21), 2(20), 2(22), and 3(20) exhibited the strongest antibacterial activity (1.7-6.8μg/mL) against MRSA (vancomycin, MIC=0.25μg/mL). The MICs of these amphiphiles against six clinical MRSA were similar to those against the ATCC strain. In PBS, EC(10)s of the most active homologues ranged from 7 to 18μg/mL and 18 to 220μg/mL for di- and tricarboxyl dendritic amphiphiles, respectively. To assess the potential safety of using dendritic amphiphiles as drugs, measurements of micellar and hemolytic properties were conducted in the same medium (full-strength BHIB) that was used for antibacterial activity. The CMCs (9-36μg/mL, ∼18-72μM) of ten amphiphiles were measured by microdilution (log2 progression) with dye-covered beads. The EC(10)s were similar to those in PBS. The MICs of most amphiphiles (14-72μg/mL) and vancomycin (1.1-2.2μg/mL) against both S. aureus and MRSA increased significantly compared to the MICs measured in 0.1-strength BHIB. The one exception, 5(18), had an MIC against S. aureus of 1.1μg/mL compared to vancomycin (2.2μg/mL). With CMC (9-18μg/mL) and EC(10) (16μg/mL) values higher than the MIC, 5(18) was discovered as a lead for further development.
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Dodecanoic acid, ≥99% (GC/titration)
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Dodecanoic acid, 98%
Supelco
Stearic acid, analytical standard
Supelco
Dodecanoic acid, analytical standard
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Supelco
Myristic acid, analytical standard
Sigma-Aldrich
Myristic acid, ≥95%, FCC, FG
Sigma-Aldrich
Myristic acid, natural, ≥98.5%, FG
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Decanoic acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Myristic acid, ≥98.0% (GC)
Supelco
Decanoic acid, analytical standard
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Stearic acid 50, tested according to Ph. Eur.
Supelco
Palmitic acid, analytical standard
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Myristic acid, Sigma Grade, ≥99%
Sigma-Aldrich
Decanoic acid, ≥98.0%
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Decanoic acid, ≥99.5%, FCC, FG