Skip to Content
Merck
All Photos(1)

Key Documents

00140585

Isoquercitrin

primary reference standard

Synonym(s):

Quercetin 3-β-D-glucoside, 3,3′,4′,5,7-Pentahydroxyflavone 3-β-glucoside, Isoquercitrin

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$787.00

$787.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$787.00

About This Item

Empirical Formula (Hill Notation):
C21H20O12
CAS Number:
Molecular Weight:
464.38
Beilstein:
100989
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$787.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

SMILES string

OC[C@H]1O[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C21H20O12/c22-6-13-15(27)17(29)18(30)21(32-13)33-20-16(28)14-11(26)4-8(23)5-12(14)31-19(20)7-1-2-9(24)10(25)3-7/h1-5,13,15,17-18,21-27,29-30H,6H2/t13-,15-,17+,18-,21+/m1/s1

InChI key

OVSQVDMCBVZWGM-QSOFNFLRSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Isoquercitrin is a flavonoid, glucoside derivative of quercitrin, and protein disulfide isomerase (PDI) inhibitor. Obtained from the leaf of medicinal plants, such as Apocynum venetum among others, it is known for its anti-oxidant and anti-thrombotic activities.
Produced and qualified by HWI pharma services GmbH.
Exact content measured by quantitative NMR can be found on the certificate.

Application

The primary reference standard can be used for identification, assay, and purity testing, method development, and validation.It can also be used as follows:
  • Development of a synergistic cloud point extraction (SCPE) method for the extraction of isoquercitrin in rat plasma samples and its subsequent analysis by high-performance liquid chromatography (HPLC)
  • Electrochemical analysis of isoquercitrin and epigallocatechin gallate by cyclic voltammetry using carbon nanotubes based electrode
  • Simultaneous determination of rutin and isoquercitrin by differential pulse voltammetry (DPV) using glassy carbon electrode (GCE) modified with deposition of thio-β-cyclodextrin functionalized graphene/palladium nanoparticles (SH-β-CD-Gr/PdNPs) for pharmaceutical and medical analysis
  • Multi-residue analysis of isoquercitrin, campesterol, emodin 8-O-β-D-glucopyranoside, and quercetin in dried stem and flower extracts of Reynoutria sachalinensis by high-performance liquid chromatography coupled with diode-array detection

Other Notes

This compound is commonly found in plants of the genus: arnica sambucus

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anti-inflammatory activity of quercetin and isoquercitrin in experimental murine allergic asthma.
Rogerio AP
Inflammation Research, 56(10), 402-408 (2007)
New insight into the flavonoid composition of Chenopodium botrys.
Bojilov, D., S. Dagnon, and I. Ivanov.
Phytochemistry Letters (2017)
Qi Chang et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 59(3), 549-555 (2005-03-12)
The present study was to investigate oral absorption of the two similar flavonoid glycosides, isoquercitrin (IQ, quercetin-3-O-glucoside) and hyperoside (HP, quercetin-3-O-galactoside) in rats. Two groups of male SD rats received an oral dose of either IQ (4.5 mg/kg) or HP
Phytochemistry Letters." New insight into the flavonoid composition of Chenopodium botrys.
Bojilov, D., S. Dagnon, and I. Ivanov.
Phytochemistry Letters (2017)
A P Rogerio et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 56(10), 402-408 (2007-11-21)
Eosinophils and cytokines are implicated in the pathogenesis of allergic diseases. In the present study, we investigate the anti-inflammatory effect of quercetin and isoquercitrin in a murine model of asthma. BALB/c mice were immunized (ovalbumin/aluminum hydroxide, s. c.), followed by

Protocols

HPLC Analysis of Polyphenols in Nero d'Avola Red Wine on Discovery® HS C18 (UV 280 nm)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service