Merck
  • Nicotinamide Treatment Facilitates Mitochondrial Fission through Drp1 Activation Mediated by SIRT1-Induced Changes in Cellular Levels of cAMP and Ca2.
Nicotinamide Treatment Facilitates Mitochondrial Fission through Drp1 Activation Mediated by SIRT1-Induced Changes in Cellular Levels of cAMP and Ca2.
Cells (2021-04-04)
Seon Beom Song, Jin Sung Park, So Young Jang, Eun Seong Hwang
ABSTRACT
Mitochondrial autophagy (or mitophagy) is essential for mitochondrial quality control, which is critical for cellular and organismal health by attenuating reactive oxygen species generation and maintaining bioenergy homeostasis. Previously, we showed that mitophagy is activated in human cells through SIRT1 activation upon treatment of nicotinamide (NAM). Further, mitochondria are maintained as short fragments in the treated cells. In the current study, molecular pathways for NAM-induced mitochondrial fragmentation were sought. NAM treatment induced mitochondrial fission, at least in part by activating dynamin-1-like protein (Drp1), and this was through attenuation of the inhibitory phosphorylation at serine 637 (S637) of Drp1. This Drp1 hypo-phosphorylation was attributed to SIRT1-mediated activation of AMP-activated protein kinase (AMPK), which in turn induced a decrease in cellular levels of cyclic AMP (cAMP) and protein kinase A (PKA) activity, a kinase targeting S637 of Drp1. Furthermore, in NAM-treated cells, cytosolic Ca2+ was highly maintained; and, as a consequence, activity of calcineurin, a Drp1-dephosphorylating phosphatase, is expected to be elevated. These results suggest that NAD+-mediated SIRT1 activation facilitates mitochondrial fission through activation of Drp1 by suppressing its phosphorylation and accelerating its dephosphorylation. Additionally, it is suggested that there is a cycle of mitochondrial fragmentation and cytosolic Ca2+-mediated Drp1 dephosphorylation that may drive sustained mitochondrial fragmentation.
Sigma-Aldrich
FK-506 monohydrate, ≥98% (HPLC)
Sigma-Aldrich
EX-527, ≥98% (HPLC)
Sigma-Aldrich
Ionomycin from Streptomyces conglobatus, ≥98% (HPLC)
Sigma-Aldrich
Sirtinol, ≥95% (HPLC)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Fluo-3, suitable for fluorescence, ~70%