Merck
  • Systematic Activity Maturation of a Single-Domain Antibody with Non-canonical Amino Acids through Chemical Mutagenesis.
Systematic Activity Maturation of a Single-Domain Antibody with Non-canonical Amino Acids through Chemical Mutagenesis.
Cell chemical biology (2020-11-21)
Philip R Lindstedt, Francesco A Aprile, Pietro Sormanni, Robertinah Rakoto, Christopher M Dobson, Gonçalo J L Bernardes, Michele Vendruscolo
ABSTRACT
Great advances have been made over the last four decades in therapeutic and diagnostic applications of antibodies. The activity maturation of antibody candidates, however, remains a significant challenge. To address this problem, we present a method that enables the systematic enhancement of the activity of a single-domain antibody through the post-translational installation of non-canonical side chains by chemical mutagenesis. We illustrate this approach by performing a structure-activity relationship study beyond the 20 naturally occurring amino acids on a single-domain antibody designed in silico to inhibit the aggregation of the amyloid-β peptide, a process closely linked to Alzheimer's disease. We found that this approach can improve, by five orders of magnitude, the anti-aggregation activity of the starting single-domain antibody, without affecting its stability. These results show that the expansion of the chemical space available to antibodies through chemical mutagenesis can be exploited for the systematic enhancement of the activity of these molecules.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Ampicillin sodium salt
Sigma-Aldrich
1-Thioglycerol, BioXtra, ≥97%
Sigma-Aldrich
Nitrilotriacetic acid disodium salt, Sigma Grade, ≥99%
Sigma-Aldrich
Guanidine hydrochloride, for molecular biology, ≥99%
Sigma-Aldrich
2-Methyl-2-propanethiol, 99%
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
N-Acetylcysteamine, 95%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Zinc, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
Acetonitrile, for UHPLC, suitable for mass spectrometry (MS)
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Cysteamine, ~95%
Sigma-Aldrich
Biocytin, ≥98% (TLC)
Sigma-Aldrich
Ammonium acetate, ≥98%
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Imidazole, ≥99% (titration)
Sigma-Aldrich
Thioflavin T, used as stain for amyloid
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Cysteamine, ≥98.0% (RT)
Sigma-Aldrich
Piperidine, ReagentPlus®, 99%
Sigma-Aldrich
1-Hexanethiol, 95%
Sigma-Aldrich
Potassium thioacetate, 98%
Sigma-Aldrich
2,5-Dibromohexanediamide, ≥95%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%