Merck
Cathepsin B is an executioner of ferroptosis.
Biochimica et biophysica acta. Molecular cell research (2020-12-20)
Pandian Nagakannan, Md Imamul Islam, Marcus Conrad, Eftekhar Eftekharpour
ABSTRACT
Ferroptosis is a necrotic form of cell death caused by inactivation of the glutathione system and uncontrolled iron-mediated lipid peroxidation. Increasing evidence implicates ferroptosis in a wide range of diseases from neurotrauma to cancer, highlighting the importance of identifying an executioner system that can be exploited for clinical applications. In this study, using pharmacological and genetic models of ferroptosis, we observed that lysosomal membrane permeabilization and cytoplasmic leakage of cathepsin B unleashes structural and functional changes in mitochondria and promotes a not previously reported cleavage of histone H3. Inhibition of cathepsin-B robustly rescued cellular membrane integrity and chromatin degradation. We show that these protective effects are independent of glutathione peroxidase-4 and are mediated by preventing lysosomal membrane damage. This was further confirmed when cathepsin B knockout primary fibroblasts remained unaffected in response to various ferroptosis inducers. Our work identifies new and yet-unrecognized aspects of ferroptosis and identifies cathepsin B as a mediator of ferroptotic cell death.
Sigma-Aldrich
CA-074 methyl ester, ≥99% (TLC)
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
Monochlorobimane, suitable for fluorescence, ≥70.0% (HPCE)
Sigma-Aldrich
L-Buthionine-sulfoximine, ≥97% (TLC)
Sigma-Aldrich
1,1,3,3-Tetramethoxypropane, 99%
Sigma-Aldrich
Erastin, ≥98% (HPLC)
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
Sigma-Aldrich
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-methylcoumarin, ≥97% (HPLC), powder
Sigma-Aldrich
L-Glutamic acid monosodium salt hydrate, ≥99% (HPLC), powder
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
Chlorobimane