Merck
  • Characterization of basal and estrogen-regulated antisense transcription in breast cancer cells: Role in regulating sense transcription.
Characterization of basal and estrogen-regulated antisense transcription in breast cancer cells: Role in regulating sense transcription.
Molecular and cellular endocrinology (2020-02-09)
Tim Y Hou, Tulip Nandu, Rui Li, Minho Chae, Shino Murakami, W Lee Kraus
ABSTRACT
Estrogen-responsive breast cancer cells exhibit both basal and estrogen-regulated transcriptional programs, which lead to the transcription of many different transcription units (i.e., genes), including those that produce coding and non-coding sense (e.g., mRNA, lncRNA) and antisense (i.e., asRNA) transcripts. We have previously characterized the global basal and estrogen-regulated transcriptomes in estrogen receptor alpha (ERα)-positive MCF-7 breast cancer cells. Herein, we have mined genomic data to define three classes of antisense transcription in MCF-7 cells based on where their antisense transcription termination sites reside relative to their cognate sense mRNA and lncRNA genes. These three classes differ in their response to estrogen treatment, the enrichment of a number of genomic features associated with active promoters (H3K4me3, RNA polymerase II, open chromatin architecture), and the biological functions of their cognate sense genes as analyzed by DAVID gene ontology. We further characterized two estrogen-regulated antisense transcripts arising from the MYC gene in MCF-7 cells, showing that these antisense transcripts are 5'-capped, 3'-polyadenylated, and localized to different compartments of the cell. Together, our analyses have revealed distinct classes of antisense transcription correlated to different biological processes and response to estrogen stimulation, uncovering another layer of hormone-regulated gene regulation.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Insulin from bovine pancreas, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and 15 mM HEPES, without sodium bicarbonate and phenol red, powder, suitable for cell culture
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Minimum Essential Medium Eagle, with Earle′s salts and non-essential amino acids, without L-glutamine, phenol red and sodium bicarbonate, Modified, powder, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, With Hanks′ salts, L-glutamine and non-essential amino acids, without sodium bicarbonate, powder, suitable for cell culture