Merck
  • Genetically encoded betaxanthin-based small-molecular fluorescent reporter for mammalian cells.
Genetically encoded betaxanthin-based small-molecular fluorescent reporter for mammalian cells.
Nucleic acids research (2020-05-19)
Pascal Stücheli, Simon Sieber, David W Fuchs, Leo Scheller, Tobias Strittmatter, Pratik Saxena, Karl Gademann, Martin Fussenegger
ABSTRACT
We designed and engineered a dye production cassette encoding a heterologous pathway, including human tyrosine hydroxylase and Amanita muscaria 4,5-DOPA dioxygenase, for the biosynthesis of the betaxanthin family of plant and fungal pigments in mammalian cells. The system does not impair cell viability, and can be used as a non-protein reporter system to directly visualize the dynamics of gene expression by profiling absorbance or fluorescence in the supernatant of cell cultures, as well as for fluorescence labeling of individual cells. Pigment profiling can also be multiplexed with reporter proteins such as mCherry or the human model glycoprotein SEAP (secreted alkaline phosphatase). Furthermore, absorbance measurement with a smartphone camera using standard application software enables inexpensive, low-tech reporter quantification.
Sigma-Aldrich
Resazurin sodium salt, powder, BioReagent
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, sterile-filtered, suitable for cell culture