Merck
  • Potent and selective steroidal inhibitors of 17beta-hydroxysteroid dehydrogenase type 7, an enzyme that catalyzes the reduction of the key hormones estrone and dihydrotestosterone.
Potent and selective steroidal inhibitors of 17beta-hydroxysteroid dehydrogenase type 7, an enzyme that catalyzes the reduction of the key hormones estrone and dihydrotestosterone.
Journal of medicinal chemistry (2009-09-24)
Edith Bellavance, Van Luu-The, Donald Poirier
ABSTRACT
17beta-Hydroxysteroid dehydrogenase type 7 (17beta-HSD7) catalyzes the reduction of estrone (E(1)) into estradiol (E(2)) and of dihydrotestosterone (DHT) into 5alpha-androstane-3beta,17beta-diol (3beta-diol), therefore modulating the level of mitogenic estrogens and androgens in humans. By classical and parallel chemistry, we generated several 4-methyl-4-aza-5alpha-androstane derivatives differing in their C-17 substituent: 17beta-formamide, 17beta-benzamide, and 17beta-tertiary amine. Best candidates in each category had demonstrated good inhibitory potency toward the conversion of E(1) into E(2) (IC(50) = 189-451 nM) and also toward the conversion of DHT into 3beta-diol (69-91% at 3 microM). Inhibition assays with 17beta-HSD1, 17beta-HSD5, 5alpha-reductase (5alpha-R) 1 and 5alpha-R2 revealed that 17beta-HSD7 inhibitors with a 4-methyl-4-aza nucleus were also able to inhibit 5alpha-Rs but not the other enzymes tested. Two 4-aza-5alpha-androstane inhibitors were, however, selective and still showed good inhibition of 17beta-HSD7. First selective and efficient inhibitors of 17beta-HSD7 are now available for additional mechanistic and therapeutic studies.
Product Description
Supelco
Androsterone, VETRANAL®, analytical standard
Sigma-Aldrich
Coumestrol, ≥95.0% (HPLC)
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Reichstein′s substance S, ≥98%
Sigma-Aldrich
Hydrocortisone-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Corticosterone, ≥92%
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
5α-Androstane
Sigma-Aldrich
17α-Ethynylestradiol, ≥98%
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
Estrone, ≥99%
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Supelco
Progesterone, VETRANAL®, analytical standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Supelco
α-Estradiol, VETRANAL®, analytical standard
Sigma-Aldrich
Chrysin, ≥96.5%
Sigma-Aldrich
Estriol, ≥97%
Sigma-Aldrich
5-Pregnen-3β-ol-20-one, ≥98%
Sigma-Aldrich
Estriol, meets USP testing specifications
Sigma-Aldrich
17α-Hydroxyprogesterone, ≥95%