Merck
  • Relationship between brain tissue partitioning and microemulsion retention factors of CNS drugs.
Relationship between brain tissue partitioning and microemulsion retention factors of CNS drugs.
Journal of medicinal chemistry (2009-03-05)
Hong Wan, Madeleine Ahman, Anders G Holmén
ABSTRACT
In CNS drug discovery, knowledge of drug-tissue binding is essential for a better understanding of brain penetration by assessing unbound brain to plasma ratio as well as pharmacokinetics (PK) and pharmacodynamics (PD) relationship by relating free drug concentration to pharmacological effect in target tissues. In this work, we present a novel microemulsion based capillary electrophoresis (CE) method that enables coupling microemulsion electrokinetic chromatography (MEEKC) to mass spectrometry (MS) for prediction of biopartitioning of CNS drugs in brain tissue. Compared to LC retention based lipophilicity and calculated lipophilicity, a significantly improved correlation between the LogP values obtained from MEEKC retention factors and fraction unbound (fu) in brain tissue was observed for a training set of structurally diverse CNS drugs as well as for a test set of new chemical entities (NCEs). The current online CE/MS/MEEKC technique can also be a potential approach for lipophilicity screening amenable for highly predictive of other ADME-Tox properties of NCEs using the MEEKC partitioning coefficient as a relevant descriptor.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Supelco
(−)-Nicotine solution, 1.0 mg/mL, analytical standard, for drug analysis
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Supelco
Mebendazole, analytical standard, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Corticosterone, ≥92%
Sigma-Aldrich
Benzocaine, ≥99% (HPLC)
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Sigma-Aldrich
Pindolol, ≥98% (TLC), powder
Supelco
(−)-Nicotine, PESTANAL®, analytical standard
Supelco
Mebendazole, VETRANAL®, analytical standard
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Sigma-Aldrich
Piroxicam, meets USP testing specifications
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Ethyl 4-aminobenzoate, 98%