Merck
  • Antitubercular dihydroagarofuranoid sesquiterpenes from the roots of Microtropis fokienensis.
Antitubercular dihydroagarofuranoid sesquiterpenes from the roots of Microtropis fokienensis.
Journal of natural products (2007-02-24)
Jih-Jung Chen, Tsung-Hsien Chou, Chien-Fang Peng, Ih-Sheng Chen, Sheng-Zehn Yang
ABSTRACT
Four new dihydroagarofuranoid sesquiterpenes (1-4) and a new hydroxybenzylsalicylaldehyde, forkienin (5), together with nine known compounds have been isolated from the roots of Microtropis fokienensis. The structures of the new compounds were determined through analyses of physical data. Compounds 3, 4, 7, and 8 exhibited potent antitubercular activities (MICs < or = 26.0 microM) against Mycobacterium tuberculosis 90-221387 in vitro.
Product Description
Supelco
β-Sitosterol, certified reference material, 100 μg/mL in chloroform
Sigma-Aldrich
β-Sitosterol, ≥70%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Syringic acid, ≥95% (HPLC)
Sigma-Aldrich
β-Sitosterol, from soybean, ≥96%
Sigma-Aldrich
β-Sitosterol, synthetic, ≥95%
Sigma-Aldrich
4-Hydroxybenzaldehyde, ≥97%, FG
Sigma-Aldrich
Vanillic acid, 97%
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
4-Hydroxybenzaldehyde, 98%
Sigma-Aldrich
4-Hydroxybenzaldehyde, ≥95.0% (HPLC)
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)