Merck
  • Carbonic anhydrase inhibitors. Inhibition of the transmembrane isozyme XII with sulfonamides-a new target for the design of antitumor and antiglaucoma drugs?
Carbonic anhydrase inhibitors. Inhibition of the transmembrane isozyme XII with sulfonamides-a new target for the design of antitumor and antiglaucoma drugs?
Bioorganic & medicinal chemistry letters (2005-02-03)
Daniela Vullo, Alessio Innocenti, Isao Nishimori, Jaromír Pastorek, Andrea Scozzafava, Silvia Pastoreková, Claudiu T Supuran
ABSTRACT
The inhibition of a newly cloned human carbonic anhydrase (CA, EC 4.2.1.1), isozyme XII (hCA XII), has been investigated with a series of sulfonamides, including some clinically used derivatives (acetazolamide, methazolamide, ethoxzolamide, dichlorophenamide, dorzolamide, brinzolamide, benzolamide, and sulpiride, or indisulam, a compound in clinical development as antitumor drug), as well as the sulfamate antiepileptic drug topiramate. Some simple amino-/hydrazine-/hydroxy-substituted aromatic/heterocyclic sulfonamides have also been included in the study. All types of activity have been detected, with several medium potency inhibitors (K(I)s in the range of 34-220 nM), whereas ethoxzolamide and several halogenated sulfanilamides showed stronger potency, with K(I)s in the range of 11-22 nM. The antiglaucoma sulfonamides used clinically, except dichlorophenamide, which is a moderate inhibitor (K(I) of 50 nM), as well as topiramate, indisulam, and sulpiride behave as very potent hCA XII inhibitors, with K(I)s in the range of 3.0-5.7 nM. Several subnanomolar inhibitors (K(I)s in the range of 0.30-0.85 nM) have also been detected. Compounds with excellent selectivity against hCA XII over hCA II have been found, showing selectivity ratios in the range of 177.7-566.7. Apparently, hCA XII is a target of the antiglaucoma sulfonamides, and potent hCA XII inhibitors may be developed/used for the management of hypoxic tumors, together with inhibitors of the other tumor-associated isozyme, CA IX.
Sigma-Aldrich
4-Sulfamoylbenzoic acid, 97%
Sigma-Aldrich
p-Toluenesulfonamide, reagent grade, 97%
Sigma-Aldrich
p-Toluenesulfonamide, ReagentPlus®, ≥99%