Merck
  • Clearance of activity-evoked K+ transients and associated glia cell swelling occur independently of AQP4: A study with an isoform-selective AQP4 inhibitor.
Clearance of activity-evoked K+ transients and associated glia cell swelling occur independently of AQP4: A study with an isoform-selective AQP4 inhibitor.
Glia (2020-06-09)
Trine Lisberg Toft-Bertelsen, Brian Roland Larsen, Sofie Kjellerup Christensen, Himanshu Khandelia, Helle S Waagepetersen, Nanna MacAulay
ABSTRACT
The mammalian brain consists of 80% water, which is continuously shifted between different compartments and cellular structures by mechanisms that are, to a large extent, unresolved. Aquaporin 4 (AQP4) is abundantly expressed in glia and ependymal cells of the mammalian brain and has been proposed to act as a gatekeeper for brain water dynamics, predominantly based on studies utilizing AQP4-deficient mice. However, these mice have a range of secondary effects due to the gene deletion. An efficient and selective AQP4 inhibitor has thus been sorely needed to validate the results obtained in the AQP4-/- mice to quantify the contribution of AQP4 to brain fluid dynamics. In AQP4-expressing Xenopus laevis oocytes monitored by a high-resolution volume recording system, we here demonstrate that the compound TGN-020 is such a selective AQP4 inhibitor. TGN-020 targets the tested species of AQP4 with an IC50 of ~3.5 μM, but displays no inhibitory effect on the other AQPs (AQP1-AQP9). With this tool, we employed rat hippocampal slices and ion-sensitive microelectrodes to determine the role of AQP4 in glia cell swelling following neuronal activity. TGN-020-mediated inhibition of AQP4 did not prevent stimulus-induced extracellular space shrinkage, nor did it slow clearance of the activity-evoked K+ transient. These data, obtained with a verified isoform-selective AQP4 inhibitor, indicate that AQP4 is not required for the astrocytic contribution to the K+ clearance or the associated extracellular space shrinkage.
Product Description
Supelco
Potassium tetrakis(4-chlorophenyl)borate, Selectophore, ≥98.0%
Sigma-Aldrich
Ethyl 3-aminobenzoate, 97%
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥96% (HPLC), powder
Sigma-Aldrich
2-Bromo-2-chloro-1,1,1-trifluoroethane, ≥99%
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, lyophilized powder, 30-60 units/mg protein (biuret), ≤0.02% glucose
Supelco
Ethyl 3-aminobenzoate methanesulfonate salt, analytical standard
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium, Without glucose, L-glutamine, phenol red, sodium pyruvate and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type XV, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
TGN-020, ≥98% (HPLC)