Merck
  • On the synergistic catalytic properties of bimetallic mesoporous materials containing aluminum and zirconium: the Prins cyclisation of citronellal.
On the synergistic catalytic properties of bimetallic mesoporous materials containing aluminum and zirconium: the Prins cyclisation of citronellal.
Chemistry (Weinheim an der Bergstrasse, Germany) (2011-01-25)
Selvedin Telalović, Anand Ramanathan, Jeck Fei Ng, Rajamanickam Maheswari, Cees Kwakernaak, Fouad Soulimani, Hans C Brouwer, Gaik Khuan Chuah, Bert M Weckhuysen, Ulf Hanefeld
ABSTRACT
Bimetallic three-dimensional amorphous mesoporous materials, Al-Zr-TUD-1 materials, were synthesised by using a surfactant-free, one-pot procedure employing triethanolamine (TEA) as a complexing reagent. The amount of aluminium and zirconium was varied in order to study the effect of these metals on the Brønsted and Lewis acidity, as well as on the resulting catalytic activity of the material. The materials were characterised by various techniques, including elemental analysis, X-ray diffraction, high-resolution TEM, N(2) physisorption, temperature-programmed desorption (TPD) of NH(3), and (27) Al MAS NMR, XPS and FT-IR spectroscopy using pyridine and CO as probe molecules. Al-Zr-TUD-1 materials are mesoporous with surface areas ranging from 700-900 m(2) g(-1), an average pore size of around 4 nm and a pore volume of around 0.70 cm(3) g(-1). The synthesised Al-Zr-TUD-1 materials were tested as catalyst materials in the Lewis acid catalysed Meerwein-Ponndorf-Verley reduction of 4-tert-butylcyclohexanone, the intermolecular Prins synthesis of nopol and in the intramolecular Prins cyclisation of citronellal. Although Al-Zr-TUD-1 catalysts possess a lower amount of acid sites than their monometallic counterparts, according to TPD of NH(3), these materials outperformed those of the monometallic Al-TUD-1 as well as Zr-TUD-1 in the Prins cyclisation of citronellal. This proves the existence of synergistic properties of Al-Zr-TUD-1. Due to the intramolecular nature of the Prins cyclisation of citronellal, the hydrophilic surface of the catalyst as well as the presence of both Brønsted and Lewis acid sites synergy could be obtained with bimetallic Al-Zr-TUD-1. Besides spectroscopic investigation of the active sites of the catalyst material a thorough testing of the catalyst in different types of reactions is crucial in identifying its specific active sites.
Sigma-Aldrich
Tetraethyl orthosilicate, 99.999% trace metals basis
Sigma-Aldrich
(R)-(+)-Citronellal, technical grade
Sigma-Aldrich
1-Propanol, anhydrous, 99.7%
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
4-tert-Butylcyclohexanone, 99%
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Supelco
1-Propanol, analytical standard
Sigma-Aldrich
Tetraethyl orthosilicate, reagent grade, 98%
Sigma-Aldrich
Tetraethyl orthosilicate, packaged for use in deposition systems
Sigma-Aldrich
1-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
1-Propanol, ≥99% (GC), purum
Sigma-Aldrich
Tetraethyl orthosilicate, ≥99.0% (GC)
Sigma-Aldrich
1-Propanol, suitable for HPLC, ≥99.9%
Supelco
(±)-Citronellal, analytical standard
Sigma-Aldrich
Triethanolamine, puriss., meets analytical specification of NF, ≥99% (GC)
Sigma-Aldrich
(±)-Citronellal, natural, ≥85%, FCC, FG
Sigma-Aldrich
tert-Butanol, anhydrous, ≥99.5%
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)
Supelco
Toluene, analytical standard
Sigma-Aldrich
Aluminum isopropoxide, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum isopropoxide, ≥98%
Sigma-Aldrich
1,3,5-Triisopropylbenzene, 95%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Triethanolamine, reagent grade, 98%
Sigma-Aldrich
Triethanolamine, puriss. p.a., ≥99% (GC)
Supelco
tert-Butanol, analytical standard
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
tert-Butanol, puriss. p.a., ACS reagent, ≥99.7% (GC)
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
tert-Butanol, TEBOL® 99, ≥99.3%