Merck
NRF2 Augments Epidermal Antioxidant Defenses and Promotes Atopy.
Journal of immunology (Baltimore, Md. : 1950) (2020-07-22)
Tatsuya Ogawa, Yosuke Ishitsuka, Yoshiyuki Nakamura, Noriko Kubota, Akimasa Saito, Yasuhiro Fujisawa, Rei Watanabe, Naoko Okiyama, Yasushi Suga, Dennis R Roop, Manabu Fujimoto
ABSTRACT
Atopic dermatitis is a chronic form of allergic contact dermatitis that is closely associated with a compromised epidermal barrier. Immunogenicity of a given electrophilic hapten after penetration of this barrier depends directly on biochemical reactions in the thiol-rich layer in the stratum granulosum. In response to electrophilic hapten, NF-erythroid 2-related factor 2 (NRF2) in keratinocytes efficiently induces the production of antioxidants. In this study, we show that the immunogenicity of a given hapten depends directly on the extent to which it induces antioxidant host defenses within the epidermal tissue. We found that allergic contact dermatitis did not develop in NRF2-deficient mice because of compromise of the epidermal innate immune responses that upregulate IL-1α. We also analyzed epidermal NRF2 in association with congenital disorders with features similar to atopic dermatitis in humans. Epidermal samples from patients with Netherton syndrome and peeling skin syndrome exhibited elevated levels of NRF2 and also elevated levels of its downstream target, small proline-rich protein 2. Taken together, these results suggest that the thiol-mediated biochemical responses in the stratum granulosum provide a critical link between defective epidermal barrier function and the development of atopy. Likewise, our results suggested that NRF2 may have a profound impact on the generation of cutaneous immunological memory.
Sigma-Aldrich
tert-Butylhydroquinone, 97%
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
TNP, ≥95% (HPLC)
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
DL-Cysteine, technical grade