Merck
  • Loss of Sirtuin 6 in osteoblast lineage cells activates osteoclasts, resulting in osteopenia.
Loss of Sirtuin 6 in osteoblast lineage cells activates osteoclasts, resulting in osteopenia.
Bone (2020-07-01)
Sung-Jin Kim, Yongxu Piao, Moon Geon Lee, A Ruem Han, Kyoungeun Kim, Chung-Ju Hwang, Jeong Taeg Seo, Seok Jun Moon
ABSTRACT
Adult bone homeostasis requires a fine-tuned balance between the activity of osteoblasts and osteoclasts. This osteoblast-osteoclast coupling is therapeutically important because it limits the efficacy of most anabolic or anti-resorptive treatments for osteoporosis. Sirtuin6 (SIRT6), a histone deacetylase, was implicated recently as an important regulator in bone homeostasis, but its in vivo function in osteoblast lineage cells remains unclear, mainly due to a lack of in vivo experiments with osteoblast lineage-specific Sirt6 knockout mice. Here, we show that Sirt6 in mature osteoblasts and/or osteocytes inhibits osteoclastogenesis via a paracrine mechanism. We found that osteoblast/osteocyte-specific Sirt6 knockout mice show reduced bone mass due to increased osteoclast formation. Mechanistically, we attribute this increased osteoclastogenesis to decreased osteoprotegerin expression in Sirt6-null osteoblasts and osteocytes. This loss of Sirt6 in osteoblasts and osteocytes does not, however, alter bone formation parameters in vivo. It does accelerate osteogenic differentiation in ex vivo culture, indicating that the osteoblast/osteocyte-autonomous functions of SIRT6 have minor effects on the osteopenic phenotype. These results establish a critical role for SIRT6 in mature osteoblasts and osteocytes in adult bone homeostasis as a negative paracrine regulator of osteoclastogenesis.
Sigma-Aldrich
β-Glycerophosphate disodium salt hydrate, BioUltra, suitable for cell culture, suitable for plant cell culture, ≥99% (titration)
Sigma-Aldrich
Alizarin Red S, certified by the BSC
Sigma-Aldrich
Alizarin-3-methyliminodiacetic acid
Sigma-Aldrich
BCIP®/NBT Liquid Substrate System, ready to use solution
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Vivaspin 20 centrifugal concentrator, MWCO 100 kDa
Sigma-Aldrich
Alkaline Phosphatase Yellow (pNPP) Liquid Substrate System for ELISA, ready to use solution