Merck
  • RasGRP1 is a causal factor in the development of l-DOPA-induced dyskinesia in Parkinson's disease.
RasGRP1 is a causal factor in the development of l-DOPA-induced dyskinesia in Parkinson's disease.
Science advances (2020-05-20)
Mehdi Eshraghi, Uri Nimrod Ramírez-Jarquín, Neelam Shahani, Tommaso Nuzzo, Arianna De Rosa, Supriya Swarnkar, Nicole Galli, Oscar Rivera, George Tsaprailis, Catherina Scharager-Tapia, Gogce Crynen, Qin Li, Marie-Laure Thiolat, Erwan Bezard, Alessandro Usiello, Srinivasa Subramaniam
ABSTRACT
The therapeutic effects of l-3,4-dihydroxyphenylalanine (l-DOPA) in patients with Parkinson's disease (PD) severely diminishes with the onset of abnormal involuntary movement, l-DOPA-induced dyskinesia (LID). However, the molecular mechanisms that promote LID remain unclear. Here, we demonstrated that RasGRP1 [(guanine nucleotide exchange factor (GEF)] controls the development of LID. l-DOPA treatment rapidly up-regulated RasGRP1 in the striatum of mouse and macaque model of PD. The lack of RasGRP1 in mice (RasGRP1-/- ) dramatically diminished LID without interfering with the therapeutic effects of l-DOPA. Besides acting as a GEF for Ras homolog enriched in the brain (Rheb), the activator of the mammalian target of rapamycin kinase (mTOR), RasGRP1 promotes l-DOPA-induced extracellular signal-regulated kinase (ERK) and the mTOR signaling in the striatum. High-resolution tandem mass spectrometry analysis revealed multiple RasGRP1 downstream targets linked to LID vulnerability. Collectively, the study demonstrated that RasGRP1 is a critical striatal regulator of LID.
Sigma-Aldrich
Anti-Huntingtin Protein Antibody, a.a. 181-810, clone 1HU-4C8, ascites fluid, clone 1HU-4C8, Chemicon®
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, clone LNC1, ascites fluid, clone LNC1, Chemicon®
Sigma-Aldrich
Anti-GAD 65/67, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-RasGRP1 Antibody, clone 10.1, clone 10.1, from mouse
Sigma-Aldrich
Tetraethylammonium borohydride, technical, ≥95% (T)
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and 15 mM HEPES, without sodium bicarbonate and phenol red, powder, suitable for cell culture
SAFC
DMEM/Nutrient Mixture F-12 Ham, With 15 mM HEPES, without L-glutamine, L-leucine, L-lysine, L-methionine, CaCl2, MgCl2, MgSO4, sodium bicarbonate, and phenol red, powder, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human CYTH3