Merck
  • Kisspeptin-GPR54 signaling in mouse NO-synthesizing neurons participates in the hypothalamic control of ovulation.
Kisspeptin-GPR54 signaling in mouse NO-synthesizing neurons participates in the hypothalamic control of ovulation.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2012-01-21)
Naresh Kumar Hanchate, Jyoti Parkash, Nicole Bellefontaine, Danièle Mazur, William H Colledge, Xavier d'Anglemont de Tassigny, Vincent Prevot
ABSTRACT
Reproduction is controlled in the brain by a neural network that drives the secretion of gonadotropin-releasing hormone (GnRH). Various permissive homeostatic signals must be integrated to achieve ovulation in mammals. However, the neural events controlling the timely activation of GnRH neurons are not completely understood. Here we show that kisspeptin, a potent activator of GnRH neuronal activity, directly communicates with neurons that synthesize the gaseous transmitter nitric oxide (NO) in the preoptic region to coordinate the progression of the ovarian cycle. Using a transgenic Gpr54-null IRES-LacZ knock-in mouse model, we demonstrate that neurons containing neuronal NO synthase (nNOS), which are morphologically associated with kisspeptin fibers, express the kisspeptin receptor GPR54 in the preoptic region, but not in the tuberal region of the hypothalamus. The activation of kisspeptin signaling in preoptic neurons promotes the activation of nNOS through its phosphorylation on serine 1412 via the AKT pathway and mimics the positive feedback effects of estrogens. Finally, we show that while NO release restrains the reproductive axis at stages of the ovarian cycle during which estrogens exert their inhibitory feedback, it is required for the kisspeptin-dependent preovulatory activation of GnRH neurons. Thus, interactions between kisspeptin and nNOS neurons may play a central role in regulating the hypothalamic-pituitary-gonadal axis in vivo.
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
β-Estradiol 3-benzoate, ≥97%
Sigma-Aldrich
KISS-1 58-65
Sigma-Aldrich
KISS 107-121
Sigma-Aldrich
Metastin, KISS-1 68-121, ≥95% (HPLC)
Sigma-Aldrich
U0126, U0126, CAS 109511-58-2, is a potent and specific inhibitor of MEK1 (IC50 = 72 nM) and MEK2 (IC50 = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK.
Sigma-Aldrich
LY 294002, LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC50 = 1.4 µM). Acts on the ATP-binding site.
Sigma-Aldrich
Donkey serum
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Sesame oil, Antioxidant, delivery vehicle for fat-soluble compounds