Skip to Content
Merck
All Photos(1)

Key Documents

662005

Sigma-Aldrich

U0126

≥98% (HPLC), solid, MEK inhibitor, Calbiochem®

Synonym(s):

U0126, 1,4-Diamino-2,3-dicyano-1,4- bis(2-aminophenylthio)butadiene, MEK Inhibitor VI, 1,4-Diamino-2,3-dicyano-1,4-bis(2-aminophenylthio)butadiene, MEK Inhibitor VI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N6S2 · 0.5C2H6O
CAS Number:
Molecular Weight:
403.52
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

U0126, U0126, CAS 109511-58-2, is a potent and specific inhibitor of MEK1 (IC₅₀ = 72 nM) and MEK2 (IC₅₀ = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 200 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

S(c2c(cccc2)N)\C(=C(\C(=C(\Sc1c(cccc1)N)/N)\C#N)/C#N)\N

InChI

1S/C18H16N6S2/c19-9-11(17(23)25-15-7-3-1-5-13(15)21)12(10-20)18(24)26-16-8-4-2-6-14(16)22/h1-8H,21-24H2/b17-11+,18-12+

InChI key

DVEXZJFMOKTQEZ-JYFOCSDGSA-N

General description

A potent and specific inhibitor of MEK1 (IC50 = 72 nM) and MEK2 (IC50 = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK. Has very little effect on other kinases such as Abl, Cdk2, Cdk4, ERK, JNK, MEKK, MKK-3, MKK-4/SEK, MKK-6, PKC, and Raf. Shown to block doxazosin-induced osteoblastic differentiation. Also acts as an immunosuppressant by effectively blocking IL-2 synthesis and T cell proliferation without affecting the long-term outcomes of either T cell activation or tolerance. Noncompetitive with respect to ATP. Also available in InSolution format (Cat. No. 662009).

Biochem/physiol Actions

Cell permeable: no
Primary Target
MEK1 and MEK2
Product does not compete with ATP.
Reversible: no
Target IC50: 72 nM, 58 nm, against MEK1, and MEK2, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Unstable in solution; reconstitute just prior to use.

Other Notes

Choi, Y.J., et al. 2011. Biochem. Biophys. Res. Commun.416, 232.
DeSilva, D.R., et al. 1998. J. Immunol. 160, 4175.
Duncia, J.V., et al. 1998. Biorg. Med. Chem. Lett.8, 2839.
Favata, M.F., et al. 1998. J. Biol. Chem. 273, 18623.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gil-Ran Kim et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 8(14), 2004973-2004973 (2021-07-27)
Regulatory T cells play a key role in immune tolerance to self-antigens, thereby preventing autoimmune diseases. However, no drugs targeting Treg cells have been approved for clinical trials yet. Here, a chimeric peptide is generated by conjugation of the cytoplasmic
Shah Md Toufiqur Rahman et al.
The Journal of biological chemistry, 295(25), 8494-8504 (2020-05-07)
The selective pressure imposed by extrinsic death signals and stressors adds to the challenge of isolating and interpreting the roles of proteins in stress-activated signaling networks. By expressing a kinase with activating mutations and a caged lysine blocking the active
Jie-Ning Li et al.
Frontiers in cell and developmental biology, 9, 671244-671244 (2021-07-23)
MicroRNAs (miRNAs) are small non-coding RNAs which post-transcriptionally suppress target mRNAs expression and/or translation to modulate pathophyological processes. Expression and function of miRNAs are fine-tuned by a conserved biogenesis machinery involves two RNase-dependent processing steps of miRNA maturation and the
Beatriz Rendon-Mitchell et al.
Journal of immunology (Baltimore, Md. : 1950), 170(7), 3890-3897 (2003-03-21)
We recently discovered that a ubiquitous protein, high mobility group box 1 protein (HMGB1), is released by activated macrophages, and functions as a late mediator of lethal systemic inflammation. To elucidate mechanisms underlying the regulation of HMGB1 release, we examined
Milica Bozic et al.
Nature communications, 11(1), 1943-1943 (2020-04-25)
Kidney fibrosis is a highly deleterious process and a final manifestation of chronic kidney disease. Alpha-(α)-synuclein (SNCA) is an actin-binding neuronal protein with various functions within the brain; however, its role in other tissues is unknown. Here, we describe the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service