Merck
Evaluation of mitochondria in oocytes following γ-irradiation.
Scientific reports (2019-12-29)
Qiaochu Wang, Jessica M Stringer, Jun Liu, Karla J Hutt
ABSTRACT
Standard cytotoxic cancer treatments, such as radiation, can damage and deplete the supply of oocytes stored within the ovary, which predisposes females to infertility and premature menopause later in life. The mechanisms by which radiation induces oocyte damage have not been completely elucidated. The objective of this study was to determine if γ-irradiation changes mitochondrial characteristics in oocytes, possibly contributing to a reduction in oocyte number and quality. Immature oocytes were collected from postnatal day (PN) 9-11 C57Bl6 mice 3, 6 and 24 hours after 0.1 Gy γ-irradiation to monitor acute mitochondrial changes. Oocytes were classified as small (>20 µm) or growing (40-60 µm). Mitochondrial membrane potential was lost in 20% and 44% of small oocytes (~20 µm) at 3 and 6 hours after γ-irradiation, respectively, consistent with the induction of apoptosis. However, mitochondrial mass, distribution and membrane potential in the surviving small oocytes were similar to the non-irradiated controls at both time points. At 24 hours after γ-irradiation, all mitochondrial parameters analysed within immature oocytes were similar to untreated controls. Mitochondrial parameters within growing oocytes were also similar to untreated controls. When mice were superovulated more than 3 weeks after γ-irradiation, there was a significant reduction in the number of mature oocytes harvested compared to controls (Control 18 ± 1 vs 0.1 Gy 4 ± 1, n = 6/16 mice, p < 0.05). There was a slight reduction in mitochondrial mass in mature oocytes after γ-irradiation, though mitochondrial localization, mtDNA copy number and ATP levels were similar between groups. In summary, this study shows that γ-irradiation of pre-pubertal mice is associated with loss of mitochondrial membrane potential in a significant proportion of small immature oocytes and a reduction in the number of mature oocytes harvested from adult mice. Furthermore, these results suggest that immature oocytes that survive γ-irradiation and develop through to ovulation contain mitochondria with normal characteristics. Whether the oocytes that survive radiation and eventually undergo meiosis can support fertility remains to be determined.
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) bioluminescent somatic cell assay kit, for cellular ATP determination
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, sterile-filtered, suitable for cell culture
Sigma-Aldrich
M2 medium, With HEPES, without penicillin and streptomycin, liquid, sterile-filtered, suitable for mouse embryo cell culture