Merck
  • Nuclear BK channels regulate gene expression via the control of nuclear calcium signaling.
Nuclear BK channels regulate gene expression via the control of nuclear calcium signaling.
Nature neuroscience (2014-06-24)
Boxing Li, Wei Jie, Lianyan Huang, Peng Wei, Shuji Li, Zhengyi Luo, Allyson K Friedman, Andrea L Meredith, Ming-Hu Han, Xin-Hong Zhu, Tian-Ming Gao
ABSTRACT
Ion channels are essential for the regulation of neuronal functions. The significance of plasma membrane, mitochondrial, endoplasmic reticulum and lysosomal ion channels in the regulation of Ca(2+) is well established. In contrast, surprisingly little is known about the function of ion channels on the nuclear envelope (NE). Here we demonstrate the presence of functional large-conductance, calcium-activated potassium channels (BK channels) on the NE of rodent hippocampal neurons. Functionally, blockade of nuclear BK channels (nBK channels) induces NE-derived Ca(2+) release, nucleoplasmic Ca(2+) elevation and cyclic AMP response element binding protein (CREB)-dependent transcription. More importantly, blockade of nBK channels regulates nuclear Ca(2+)-sensitive gene expression and promotes dendritic arborization in a nuclear Ca(2+)-dependent manner. These results suggest that the nBK channel functions as a molecular link between neuronal activity and nuclear Ca(2+) to convey signals from synapse to nucleus and is a new modulator, operating at the NE, of synaptic activity-dependent neuronal functions.
Sigma-Aldrich
U0126 ethanolate, ≥98% (HPLC), powder
Sigma-Aldrich
(+)-Bicuculline, ≥97.0% (TLC)
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Anti-Slo1 Antibody, clone L6/60, clone L6/60, from mouse