Merck
  • Ultrasound-assisted one-phase solvent extraction coupled with liquid chromatography-quadrupole time-of-flight mass spectrometry for efficient profiling of egg yolk lipids.
Ultrasound-assisted one-phase solvent extraction coupled with liquid chromatography-quadrupole time-of-flight mass spectrometry for efficient profiling of egg yolk lipids.
Food chemistry (2020-03-18)
Ya Xie, Bangfu Wu, Zongyuan Wu, Xinghao Tu, Shuling Xu, Xin Lv, Hongqing Yin, Jiqian Xiang, Hong Chen, Fang Wei
ABSTRACT
In this research, a novel ultrasound-assisted extraction (USAE) method using a one-phase solvent (isopropanol, IPA) combined with liquid chromatography-electrospray ionization quadrupole time-of-flight mass spectrometry (LC-ESI-QToF-MS) was developed for exhaustive profiling of egg yolk lipids, which are considered to be essential nutrients that have important functions for human health. Compared with IPA-vortex extraction method, the optimized IPA-USAE method shows better performance in terms of extraction time, lipid losses, lipid recoveries and the number of lipids identified. With this established simple, high throughput and reliable analytical approach, untargeted comprehensive lipid profiling of four kinds of egg yolks was achieved, thus providing a powerful tool for nutritional research of egg lipids. Considering that the lipids contained in egg yolk are quite abundant in both quantity and species, the proposed IPA-USAE combined with LC-ESI-QToF-MS is also potentially applicable to comprehensive extraction and profiling of lipids in other biological samples.
Product Description
Avanti
15:0-18:1-d7-15:0 TG, Avanti Research - A Croda Brand 791648C
Avanti
15:0-18:1-d7-PG, Avanti Research - A Croda Brand 791640C
Avanti
17:0 cholesteryl ester, cholest-5-en-3β-yl heptadecanoate, methylene chloride solution
Avanti
15:0-18:1-d7-PA, Avanti Research - A Croda Brand
Avanti
15:0-18:1-d7 DG, Avanti Research - A Croda Brand 791647C
Avanti
12:0 SM (d18:1/12:0), Avanti Research - A Croda Brand 860583P, powder
Avanti
C17 Ceramide (d18:1/17:0), Avanti Research - A Croda Brand