Merck
  • Effect of the SSeCKS-TRAF6 interaction on gastrodin-mediated protection against 2,3,7,8-tetrachlorodibenzo-p-dioxin-induced astrocyte activation and neuronal death.
Effect of the SSeCKS-TRAF6 interaction on gastrodin-mediated protection against 2,3,7,8-tetrachlorodibenzo-p-dioxin-induced astrocyte activation and neuronal death.
Chemosphere (2019-04-09)
Man Jiao, Kaizhi Yin, Tao Zhang, Changyue Wu, Yan Zhang, Xinyuan Zhao, Qiyun Wu
ABSTRACT
The ubiquitous environmental pollutant 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD) has been shown to trigger neurotoxicity. In this study, we investigated the protective effects of gastrodin on TCDD-induced neurotoxicity and the underlying molecular mechanisms. The results show that gastrodin decreased cell viability, tumor necrosis factor-α (TNF-α) and interleukin-6 (IL-6) release, and inducible nitrix oxide synthase (iNOS) and glial fibrillary acidic protein (GFAP) expression in TCDD-treated C6 cells. TCDD stimulated NF-κB signalling activation, demonstrated by increased p-NF-κB expression and translocation of nuclear Factor kappa B (NF-κB) to the nucleus. TCDD did not affect TRAF6 protein expression but enhanced the attenuated the Src-suppressed-C Kinase Substrate (SSeCKS)-tumor necrosis factor receptor-associated factor 6 (TRAF6) interaction, thereby triggering NF-κB signalling activation. Gastrodin inhibited TCDD-induced NF-κB signalling activation by lessening the SSeCKS-TRAF6 interaction in vitro. Gastrodin attenuated SSeCKS-TRAF6 interaction in vivo and protected mice from NF-κB signalling activation following TCDD exposure. Finally, gastrodin blocked the apoptosis of PC12 neuronal cells induced by medium conditioned with TCDD-treated astrocytes. In summary, gastrodin inhibited TCDD-induced NF-κB signalling activation by lessening the SSeCKS-TRAF6 interaction, resulting in attenuated astrocyte activation and subsequent neuronal apoptosis. These findings will contribute to an improved understanding of TCDD-induced neurotoxicity and strategies to antagonise it using gastrodin.
Sigma-Aldrich
MISSION® esiRNA, targeting human TRAF6
Sigma-Aldrich
Gastrodin, ≥98% (HPLC)