Skip to Content
Merck
All Photos(1)

Documents

EHU147511

Sigma-Aldrich

MISSION® esiRNA

targeting human TRAF6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCCCAGGCTGTTCATAGTTTGAGCGTTATACCCGACTCTGGGTATATCTCAGAGGTCCGGAATTTCCAGGAAACTATTCACCAGTTAGAGGGTCGCCTTGTAAGACAAGACCATCAAATCCGGGAGCTGACTGCTAAAATGGAAACTCAGAGTATGTATGTAAGTGAGCTCAAACGAACCATTCGAACCCTTGAGGACAAAGTTGCTGAAATCGAAGCACAGCAGTGCAATGGAATTTATATTTGGAAGATTGGCAACTTTGGAATGCATTTGAAATGTCAAGAAGAGGAGAAACCTGTTGTGATTCATAGCCCTGGATTCTACACTGGCAAACCCGGGTACAAACTGTGCATGCGCTTGCACCTTCAGTTACCGACTGCTCAGCGCTGTGCAAACTATATATCCCTTTTTGTCCACACAATGCAAGGAGAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yancan Liang et al.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology, 47(6), 583-589 (2018-03-27)
Tumor necrosis factor (TNF) receptor-associated factor 6 (TRAF6) has been proved to play an important role in tumorigenesis, invasion, and metastasis. However, its precise role salivary adenoid cystic carcinoma (SACC) has not been determined. The aim of this study was
Zhiyong He et al.
Oncology reports, 35(4), 1933-1940 (2016-02-06)
Tumor necrosis factor receptor-associated factor 6 (TRAF6) has been found to be involved in multiple cancers. However, the effect of small interfering RNA (siRNA)‑induced knockdown of TRAF6 on the biological behaviors of cancer cells remains unknown. Thus, the present study
Quanfeng Wu et al.
Cancer cell international, 17, 62-62 (2017-06-09)
To study the mechanism by which epithelial ovarian cancer (EOC)-derived exosomes restore the migration of endothelial cells that is suppressed by TAM-derived exosomes. Exosomes were isolated from TAMs in the ascites of patients with EOC. The effect of exosomes on
Mengting Yang et al.
Stem cells international, 2020, 3296192-3296192 (2020-07-30)
Gastric cancer is the third most common type of tumor associated with death. TRAF6 belongs to the tumor necrosis factor receptor-associated factor family and has been demonstrated to be involved in tumor progression in various cancers. However, the exact effect
Carrie M Rosenberger et al.
PLoS pathogens, 13(4), e1006305-e1006305 (2017-04-06)
Antiviral responses must rapidly defend against infection while minimizing inflammatory damage, but the mechanisms that regulate the magnitude of response within an infected cell are not well understood. miRNAs are small non-coding RNAs that suppress protein levels by binding target

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service