Merck
  • "Clocks" in the NAD World: NAD as a metabolic oscillator for the regulation of metabolism and aging.
"Clocks" in the NAD World: NAD as a metabolic oscillator for the regulation of metabolism and aging.
Biochimica et biophysica acta (2009-11-10)
Shin-Ichiro Imai
ABSTRACT
SIR2 (silent information regulator 2) proteins, now called "sirtuins," are an evolutionarily conserved family of NAD-dependent protein deacetylases/ADP-ribosyltransferases. Sirtuins have recently attracted major attention in the field of aging research, and it has been demonstrated that SIR2 and its orthologs regulate aging and longevity in yeast, worms, and flies. In mammals, the SIR2 ortholog SIRT1 coordinates important metabolic responses to nutritional availability in multiple tissues. Most recently, it has been demonstrated that SIRT1 regulates the amplitude and the duration of circadian gene expression through the interaction and the deacetylation of key circadian clock regulators, such as BMAL1 and PER2. More strikingly, we and others have discovered a novel circadian clock feedback loop in which both the rate-limiting enzyme in mammalian NAD biosynthesis, nicotinamide phosphoribosyltransferase (NAMPT), and NAD levels display circadian oscillations and modulate CLOCK:BMAL1-mediated circadian transcriptional regulation through SIRT1, demonstrating a new function of NAD as a "metabolic oscillator." These findings reveal a novel system dynamics of a recently proposed systemic regulatory network regulated by NAMPT-mediated NAD biosynthesis and SIRT1, namely, the NAD World. In the light of this concept, a new connection between physiological rhythmicity, metabolism, and aging will be discussed.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate, ~98%, pkg of 100 mg
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate, ≥94% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate, ≥97% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide sodium salt
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt, ~98%, pkg of 5 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt, ~98%, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt, ~98%, pkg of 0.2 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt, ~98%, pkg of 25-30 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt, ~98%, pkg of 2 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced dipotassium salt
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%