Merck
  • Pseudomonas aeruginosa utilizes host polyunsaturated phosphatidylethanolamines to trigger theft-ferroptosis in bronchial epithelium.
Pseudomonas aeruginosa utilizes host polyunsaturated phosphatidylethanolamines to trigger theft-ferroptosis in bronchial epithelium.
The Journal of clinical investigation (2018-09-11)
Haider H Dar, Yulia Y Tyurina, Karolina Mikulska-Ruminska, Indira Shrivastava, Hsiu-Chi Ting, Vladimir A Tyurin, James Krieger, Claudette M St Croix, Simon Watkins, Erkan Bayir, Gaowei Mao, Catherine R Armbruster, Alexandr Kapralov, Hong Wang, Matthew R Parsek, Tamil S Anthonymuthu, Abiola F Ogunsola, Becca A Flitter, Cody J Freedman, Jordan R Gaston, Theodore R Holman, Joseph M Pilewski, Joel S Greenberger, Rama K Mallampalli, Yohei Doi, Janet S Lee, Ivet Bahar, Jennifer M Bomberger, Hülya Bayır, Valerian E Kagan
ABSTRACT
Ferroptosis is a death program executed via selective oxidation of arachidonic acid-phosphatidylethanolamines (AA-PE) by 15-lipoxygenases. In mammalian cells and tissues, ferroptosis has been pathogenically associated with brain, kidney, and liver injury/diseases. We discovered that a prokaryotic bacterium, Pseudomonas aeruginosa, that does not contain AA-PE can express lipoxygenase (pLoxA), oxidize host AA-PE to 15-hydroperoxy-AA-PE (15-HOO-AA-PE), and trigger ferroptosis in human bronchial epithelial cells. Induction of ferroptosis by clinical P. aeruginosa isolates from patients with persistent lower respiratory tract infections was dependent on the level and enzymatic activity of pLoxA. Redox phospholipidomics revealed elevated levels of oxidized AA-PE in airway tissues from patients with cystic fibrosis (CF) but not with emphysema or CF without P. aeruginosa. We believe that the evolutionarily conserved mechanism of pLoxA-driven ferroptosis may represent a potential therapeutic target against P. aeruginosa-associated diseases such as CF and persistent lower respiratory tract infections.
Sigma-Aldrich
PD 146176, ≥98% (HPLC), solid
Sigma-Aldrich
Anti-Goat IgG (whole molecule)–Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
L-(+)-Arabinose, ≥99% (GC)
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
MISSION® esiRNA, targeting human LPCAT3
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Supelco
LB broth, Miller, Microbiologically tested
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Ferrostatin-1, ≥95% (HPLC)