Merck
  • The Mevalonate Pathway Is a Druggable Target for Vaccine Adjuvant Discovery.
The Mevalonate Pathway Is a Druggable Target for Vaccine Adjuvant Discovery.
Cell (2018-10-03)
Yun Xia, Yonghua Xie, Zhengsen Yu, Hongying Xiao, Guimei Jiang, Xiaoying Zhou, Yunyun Yang, Xin Li, Meng Zhao, Liping Li, Mingke Zheng, Shuai Han, Zhaoyun Zong, Xianbin Meng, Haiteng Deng, Huahu Ye, Yunzhi Fa, Haitao Wu, Eric Oldfield, Xiaoyu Hu, Wanli Liu, Yan Shi, Yonghui Zhang
ABSTRACT
Motivated by the clinical observation that interruption of the mevalonate pathway stimulates immune responses, we hypothesized that this pathway may function as a druggable target for vaccine adjuvant discovery. We found that lipophilic statin drugs and rationally designed bisphosphonates that target three distinct enzymes in the mevalonate pathway have potent adjuvant activities in mice and cynomolgus monkeys. These inhibitors function independently of conventional "danger sensing." Instead, they inhibit the geranylgeranylation of small GTPases, including Rab5 in antigen-presenting cells, resulting in arrested endosomal maturation, prolonged antigen retention, enhanced antigen presentation, and T cell activation. Additionally, inhibiting the mevalonate pathway enhances antigen-specific anti-tumor immunity, inducing both Th1 and cytolytic T cell responses. As demonstrated in multiple mouse cancer models, the mevalonate pathway inhibitors are robust for cancer vaccinations and synergize with anti-PD-1 antibodies. Our research thus defines the mevalonate pathway as a druggable target for vaccine adjuvants and cancer immunotherapies.
Sigma-Aldrich
Octoclothepin maleate salt, solid
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥90% (agarose gel electrophoresis)
Sigma-Aldrich
Mevastatin, ≥98% (HPLC), powder or crystals
Sigma-Aldrich
TWEEN® 20, Low-peroxide; Low-carbonyls
TH-Z93, ≥98% (HPLC)
Supelco
Squalene, analytical standard
Sigma-Aldrich
Squalene, ≥98%, liquid
Sigma-Aldrich
Pravastatin sodium salt hydrate, ≥98% (HPLC), powder
Supelco
Simvastatin, analytical standard
Sigma-Aldrich
TWEEN® 80, Molecular Biology, syrup
Sigma-Aldrich
Span® 85