Merck
  • mTOR complex 1 activity is required to maintain the canonical endocytic recycling pathway against lysosomal delivery.
mTOR complex 1 activity is required to maintain the canonical endocytic recycling pathway against lysosomal delivery.
The Journal of biological chemistry (2017-02-16)
Kristin Dauner, Walaa Eid, Riya Raghupathy, John F Presley, Xiaohui Zha
ABSTRACT
The plasma membrane of mammalian cells undergoes constitutive endocytosis, endocytic sorting, and recycling, which delivers nutrients to the lysosomes. The receptors, along with membrane lipids, are normally returned to the plasma membrane to sustain this action. It is not known, however, whether this process is influenced by metabolic conditions. Here we report that endocytic recycling requires active mechanistic target of rapamycin (aka mammalian target of rapamycin) (mTORC1), a master metabolic sensor. Upon mTORC1 inactivation, either by starvation or by inhibitor, recycling receptors and plasma membrane lipids, such as transferrin receptors and sphingomyelin, are delivered to the lysosomes. This lysosomal targeting is independent of canonical autophagy: both WT and Atg5-/- mouse embryonic fibroblasts responded similarly. Furthermore, we identify hepatocyte growth factor-regulated tyrosine kinase substrate (Hrs), an endosomal sorting complexes required for transport (ESCORT-0) component, as a downstream target of mTORC1. Hrs requires mTORC1 activity to maintain its protein expression level. Silencing Hrs without decreasing mTORC1 activity is sufficient to target transferrin and sphingomyelin to the lysosomes. It is thus evident that the canonical recycling pathway is under the regulation of mTORC1 and likely most predominant in proliferating cells where mTORC1 is highly active.
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Anti-phospho-p70 S6 Kinase (Thr389) Antibody, clone 10G7.1, clone 10G7.1, from mouse
Avanti
C6-NBD Sphingomyelin, Avanti Research - A Croda Brand 810218C
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder