Skip to Content
Merck
All Photos(1)

Documents

810218C

Avanti

C6-NBD Sphingomyelin

Avanti Research - A Croda Brand 810218C

Synonym(s):

N-[6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl]-sphingosine-1-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H61N6O9P
CAS Number:
Molecular Weight:
740.87
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810218C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810218C

concentration

1 mg/mL (810218C-1mg)

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C35H61N6O9P/c1-5-6-7-8-9-10-11-12-13-14-15-16-18-21-32(42)30(28-49-51(46,47)48-27-26-41(2,3)4)37-33(43)22-19-17-20-25-36-29-23-24-31(40(44)45)35-34(29)38-50-39-35/h18,21,23-24,30,32,42H,5-17,19-20,22,25-28H2,1-4H3,(H2-,36,37,39,43,46,47)/b21-18+/t30-,32+/m0/s1

InChI key

YDBDYTGWMRUUSN-PERJAUJZSA-N

General description

C6-NBD Sphingomyelin is a fluorescent derivative of sphingomyelin. Its fluorescent property is due to presence of NBD (2-(4-nitro-2,1,3-benzoxadiazol-7-yl) aminoethyl) group. Sphingomyelin is generally concentrated in the outer leaflet of the plasma membrane.

Application

C6-NBD Sphingomyelin is suitable to study sphingomyelin endocytosis using fluorescence microscopy.

Biochem/physiol Actions

Sphingomyelin (SM) is involved in signal transduction and membrane trafficking. It is synthesized by the transfer of phosphocholine from phosphatidylcholine onto ceramide, catalysed by sphingomyelin synthase on the cell surface and in lumen of the Golgi. SM has high packing density and affinity for sterol which offer a strong barrier to extracellular environment.

Packaging

5 mL Amber Glass Screw Cap Vial (810218C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorescent and Luminescent Probes for Biological Activity (1999)
mTOR complex 1 activity is required to maintain the canonical endocytic recycling pathway against lysosomal delivery
Dauner K, et al.
The Journal of biological chemistry, 292(14), 5737-5747 (2017)
Kristin Dauner et al.
The Journal of biological chemistry, 292(14), 5737-5747 (2017-02-16)
The plasma membrane of mammalian cells undergoes constitutive endocytosis, endocytic sorting, and recycling, which delivers nutrients to the lysosomes. The receptors, along with membrane lipids, are normally returned to the plasma membrane to sustain this action. It is not known
Mark L Schultz et al.
BMC medicine, 17(1), 200-200 (2019-11-13)
Niemann-Pick disease type C is a fatal and progressive neurodegenerative disorder characterized by the accumulation of unesterified cholesterol in late endosomes and lysosomes. We sought to develop new therapeutics for this disorder by harnessing the body's endogenous cholesterol scavenging particle
Avery L McIntosh et al.
Lipids, 45(6), 465-477 (2010-05-18)
During the last several years, intracellular lipid droplets have become the focus of intense study. No longer an inert bystander, the lipid droplet is now known as a dynamic organelle contributing lipids to many cellular events. However, while the dynamics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service