Merck
  • Compartmentalized activities of the pyruvate dehydrogenase complex sustain lipogenesis in prostate cancer.
Compartmentalized activities of the pyruvate dehydrogenase complex sustain lipogenesis in prostate cancer.
Nature genetics (2018-01-18)
Jingjing Chen, Ilaria Guccini, Diletta Di Mitri, Daniela Brina, Ajinkya Revandkar, Manuela Sarti, Emiliano Pasquini, Abdullah Alajati, Sandra Pinton, Marco Losa, Gianluca Civenni, Carlo V Catapano, Jacopo Sgrignani, Andrea Cavalli, Rocco D'Antuono, John M Asara, Andrea Morandi, Paola Chiarugi, Sara Crotti, Marco Agostini, Monica Montopoli, Ionica Masgras, Andrea Rasola, Ramon Garcia-Escudero, Nicolas Delaleu, Andrea Rinaldi, Francesco Bertoni, Johann de Bono, Arkaitz Carracedo, Andrea Alimonti
ABSTRACT
The mechanisms by which mitochondrial metabolism supports cancer anabolism remain unclear. Here, we found that genetic and pharmacological inactivation of pyruvate dehydrogenase A1 (PDHA1), a subunit of the pyruvate dehydrogenase complex (PDC), inhibits prostate cancer development in mouse and human xenograft tumor models by affecting lipid biosynthesis. Mechanistically, we show that in prostate cancer, PDC localizes in both the mitochondria and the nucleus. Whereas nuclear PDC controls the expression of sterol regulatory element-binding transcription factor (SREBF)-target genes by mediating histone acetylation, mitochondrial PDC provides cytosolic citrate for lipid synthesis in a coordinated manner, thereby sustaining anabolism. Additionally, we found that PDHA1 and the PDC activator pyruvate dehydrogenase phosphatase 1 (PDP1) are frequently amplified and overexpressed at both the gene and protein levels in prostate tumors. Together, these findings demonstrate that both mitochondrial and nuclear PDC sustain prostate tumorigenesis by controlling lipid biosynthesis, thus suggesting this complex as a potential target for cancer therapy.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
SB-204990, ≥98% (HPLC)
Sigma-Aldrich
Anti-phospho-PDHE1-A type I (Ser293) Antibody, from rabbit, purified by affinity chromatography