Skip to Content
Merck

Skip To

SML2829

SB-204990

≥98% (HPLC), ATP citrate-lyase inhibitor, powder

Synonym(s):

(+/-) (3R*,5S*)-rel-5-[6-(2,4-Dichlorophenyl)hexyl]tetrahydro-3-hydroxy-2-oxo-3-furanacetic acid, SB 201076 prodrug, SB 204990, SB-201076 prodrug, SB201076 prodrug, SB204990

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
$154.00
25 mg
Check Cart for Availability
$626.00

About This Item

Empirical Formula (Hill Notation):
C18H22Cl2O5
CAS Number:
Molecular Weight:
389.27
UNSPSC Code:
51111800
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

$154.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

SB-204990, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Clc1c(ccc(c1)Cl)CCCCCC[C@H]2OC(=O)[C@@](C2)(O)CC(=O)O

InChI

1S/C18H22Cl2O5/c19-13-8-7-12(15(20)9-13)5-3-1-2-4-6-14-10-18(24,11-16(21)22)17(23)25-14/h7-9,14,24H,1-6,10-11H2,(H,21,22)/t14-,18+/m1/s1

InChI key

YTRNLFYTHYWDAU-KDOFPFPSSA-N

Biochem/physiol Actions

ATP citrate-lyase inhibitor SB-201076 γ-lactone prodrug with cell-permeability, oral availability and in vivo hypocholesterolaemic/hypolipidaemic efficacy.
SB-204990 is a cell-permeable and orally available γ-lactone prodrug of the non-cell-permeable SB-201076 that inhibits human and rat ATP citrate-lyase (Ki =1 μM against human and rat ACL) in a predominantly competitive manner with a small but significant uncompetitive component. SB-204990 treatment is shown to suppress the rate of cellular cholesterol and fatty acid synthesis in HepG2 cultures (by 91% & 82%, respectively, with 30 μM SB-204990), and display hypocholesterolaemic and hypolipidaemic efficacy in vivo when administered in the diet (0.05-0.25%, w/w; mice, rats, dogs).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML0391S7326S7389
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

powder, ≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

room temp

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: >5 mg/mL

solubility

DMSO: ≥30 mg/mL

solubility

H2O: 10 mg/mL, clear (warmed to 60 °C)

color

white to beige

color

white to beige

color

white to off-white

color

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters