![Testosterone-2,3,4-13C3 solution 0.1 mg/mL in methanol, 99 atom % 13C, 98% (CP)](/deepweb/assets/sigmaaldrich/product/structures/300/510/02465c88-5136-4aa7-89ad-1a8a745369af/640/02465c88-5136-4aa7-89ad-1a8a745369af.png)
Testosterone-2,3,4-13C3 solution
![Testosterone-2,3,4-13C3 solution 0.1 mg/mL in methanol, 99 atom % 13C, 98% (CP)](/deepweb/assets/sigmaaldrich/product/structures/300/510/02465c88-5136-4aa7-89ad-1a8a745369af/640/02465c88-5136-4aa7-89ad-1a8a745369af.png)
Empirical Formula (Hill Notation):
13C3C16H28O2
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing