Skip to Content
Merck
All Photos(1)

Key Documents

730610

Sigma-Aldrich

Testosterone-2,3,4-13C3 solution

0.1 mg/mL in methanol, 99 atom % 13C, 98% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$350.00

$350.00


Estimated to ship onApril 06, 2025


Request more information

Select a Size

Change View
1 ML
$350.00

About This Item

Empirical Formula (Hill Notation):
13C3C16H28O2
CAS Number:
Molecular Weight:
291.40
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$350.00


Estimated to ship onApril 06, 2025


Request more information

isotopic purity

99 atom % 13C

Quality Level

Assay

98% (CP)

form

solution

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

concentration

0.1 mg/mL in methanol

technique(s)

mass spectrometry (MS): suitable

mass shift

M+3

storage temp.

−20°C

SMILES string

C[C@]12CC[C@H]3[C@@H](CCC4=[13CH][13C](=O)[13CH2]C[C@]34C)[C@@H]1CC[C@@H]2O

InChI

1S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h11,14-17,21H,3-10H2,1-2H3/t14-,15-,16-,17-,18-,19-/m0/s1/i7+1,11+1,13+1

InChI key

MUMGGOZAMZWBJJ-KRMLYXHASA-N

Application

This product may be used as an analytical standard.

Packaging

1 mL in ampule

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shu Zhang et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 965, 183-189 (2014-07-20)
A liquid chromatography-electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS) method was developed, using testosterone-d3 as a surrogate analyte, for the simultaneous quantification of goserelin and testosterone in rat plasma. According to this method, the pharmacokinetic and pharmacodynamic data were obtained from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service