Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Supelclean
™
LC-NH
2
SPE Tube
Products
Product Category
tubes (3)
Brand
Supelco (3)
pK Value
Product Line
Chemical Composition
Separation Technique
Volume (ml)
Application
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
57014
Supelclean
™
LC-NH
2
SPE Tube,
bed wt. 500 mg, volume 3 mL, pk of 54
Expand
View Pricing
54059-U
Supelclean
™
LC-NH
2
SPE Tube,
bed wt. 500 mg, volume 6 mL, pk of 30
Expand
View Pricing
504483
Supelclean
™
LC-NH
2
SPE Tube,
bed wt. 100 mg, volume 1 mL, pk of 108
Expand
View Pricing