
SANT-1

Synonym(s):
(4-Benzyl-piperazin-1-yl)-(3,5-dimethyl-1-phenyl-1H-pyrazol-4-ylmethylene)-amine
Empirical Formula (Hill Notation):
C23H27N5
CAS Number:
Molecular Weight:
373.49
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing