![N,N′-Diethylthiourea 98%](/deepweb/assets/sigmaaldrich/product/structures/707/431/0f4963e9-993a-4cf0-a2a7-da9c64c236c9/640/0f4963e9-993a-4cf0-a2a7-da9c64c236c9.png)
N,N′-Diethylthiourea
![N,N′-Diethylthiourea 98%](/deepweb/assets/sigmaaldrich/product/structures/707/431/0f4963e9-993a-4cf0-a2a7-da9c64c236c9/640/0f4963e9-993a-4cf0-a2a7-da9c64c236c9.png)
Synonym(s):
N,N′-Diethylthiourea
Linear Formula:
(C2H5NH)2CS
CAS Number:
Molecular Weight:
132.23
Products
Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing