Skip to Content
Merck
All Photos(2)

Key Documents

8.03520

Sigma-Aldrich

N,N′-Diethylthiourea

for synthesis

Synonym(s):

N,N′-Diethylthiourea, 1,3-Diethylthiourea

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5NH)2CS
CAS Number:
Molecular Weight:
132.23
MDL number:
UNSPSC Code:
12352111
EC Index Number:
203-308-5
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.0000064 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (HPLC)

form

crystals

potency

316 mg/kg LD50, oral (Rat)

pH

7 (20 °C, 38 g/L in H2O)

mp

75-78 °C

transition temp

flash point 186 °C

density

1.12 g/cm3 at 20 °C

bulk density

735 kg/m3

storage temp.

2-30°C

SMILES string

S=C(NCC)NCC

InChI

1S/C5H12N2S/c1-3-6-5(8)7-4-2/h3-4H2,1-2H3,(H2,6,7,8)

InChI key

FLVIGYVXZHLUHP-UHFFFAOYSA-N

Application


  • Solid–Liquid Phase Equilibrium and Thermodynamic Analysis of N,N′-Diethylthiourea in Different Solvent Systems: This study investigates the solid–liquid phase equilibrium of N,N′-diethylthiourea in various solvent systems, analyzing both solid-state structure and solution thermodynamics (M Yao et al., 2019).

  • Appraisal of the hydrogen atom transfer mechanism for the reaction between thiourea derivatives and OH radical: A comparative study of N,N′-dimethylthiourea and N,N′-diethylthiourea with OH radicals, highlighting the less toxic nature of diethylthiourea (KRN Serobatse, MM Kabanda, 2017).

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 75 °C
Melting range (upper value): ≤ 78 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

366.8 °F

Flash Point(C)

186 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service