Skip to Content
Merck
Guanidine hydrochloride anhydrous, free-flowing, Redi-Dri™, ≥99%

Guanidine hydrochloride

Guanidine hydrochloride anhydrous, free-flowing, Redi-Dri™, ≥99%
Synonym(s):
Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidinium chloride, Guanidine hydrochloride
Linear Formula:
NH2C(=NH)NH2 · HCl
CAS Number:
Molecular Weight:
95.53
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Technique