Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Certified sec. standard reference buffer solution
Products
Brand
Supelco (5)
Product Line
Agency
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
certified reference material (4)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
1.07202
Certified sec. standard reference buffer solution,
potassium dihydrogen phosphate/di-sodium hydrogen phosphate, pH 6.86 (25 °C), Certipur
®
Expand
View Pricing
1.07204
Certified sec. standard reference buffer solution,
potassium tetraoxalate dihydrate, directly traceable to primary SRM from NIST/PTB pH(S) = 1.67₉ (25°C) Certipur
®
Expand
View Pricing
1.07200
Certified sec. standard reference buffer solution,
potassium hydrogen phthalate, directly traceable to primary SRM from NIST/PTB pH (S) = 4.00
5
(25°C) Certipur
®
Expand
View Pricing
1.07203
Certified sec. standard reference buffer solution,
di-sodium tetraborate decahydrate, directly traceable to primary SRM from NIST/PTB pH(S) = 9.18₀ (25°C) Certipur
®
Expand
View Pricing
1.07205
Certified sec. standard reference buffer solution,
potassium dihydrogen phosphate/di-sodium hydrogen phosphate, pH 7.41 (25 °C), Certipur
®
Expand
View Pricing