Skip to Content
Merck
All Photos(1)

Documents

1.07200

Supelco

Certified sec. standard reference buffer solution

potassium hydrogen phthalate, directly traceable to primary SRM from NIST/PTB pH (S) = 4.005 (25°C) Certipur®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.25

Quality Level

storage temp.

15-25°C

General description

The certified secondary standard reference buffer solution of potassium hydrogen phthalate is gravimetrically prepared using a high-quality reagent in high-purity water. Its pH value is measured by differential potentiometry with two platinum hydrogen electrodes against primary certified reference materials from NIST & PTB. The 5-point calibration method is used for its pH qualification, in accordance with DIN 19268, against certified primary reference standards as per DIN 19266.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

The buffer solution finds its use in calibration, qualification, and monitoring of pH measuring instruments and as a control for testing other buffer solutions.

Features and Benefits

  • Available in a tamper-proof 100 mL plastic bottle in solution format
  • Subjected to detailed stability studies as per ISO Guide 35
  • Product label containing relevant information on order number, article description, lot number, expiry date and safety data, in accordance with ISO Guide 31
  • Accompanied with a detailed and batch-specific certificate of analysis (CoA) for an effective quality management system
  • Measured and qualified in ISO/IEC 17025 and ISO 17034 accredited laboratory
  • Traceable to SRMs from NIST and primary certified reference standards from PTB

Analysis Note

pH-value: 4.005 - 4.011
For complete data see the enclosed certificate

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service