Skip to Content
Products
Cart
0
AR
EN
Products
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Anthrone
Synonym(s):
9(10
H
)-Anthracenone, Anthrone
Empirical Formula (Hill Notation):
C
14
H
10
O
CAS Number:
90-44-8
Molecular Weight:
194.23
EC Number:
201-994-0
Products
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
319899
Anthrone,
ACS reagent, 97%
Expand
View Pricing
8.01461
Anthrone,
for synthesis
Expand
View Pricing
52445
Anthrone,
analytical standard
Expand
View Pricing
An unknown error has occured.
EspaƱol - ES
English - EN
Learn More