Skip to Content
Merck
All Photos(1)

Documents

8.01461

Sigma-Aldrich

Anthrone

for synthesis

Synonym(s):

Anthrone, 9,10-Dihydro-9-oxoanthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10O
CAS Number:
Molecular Weight:
194.23
MDL number:
UNSPSC Code:
12352115
EC Index Number:
201-994-0
NACRES:
NA.22

Quality Level

form

solid

mp

151-156 °C

solubility

4.6 mg/L

bulk density

450 kg/m3

storage temp.

no temp limit

InChI

1S/C14H10O/c15-14-12-7-3-1-5-10(12)9-11-6-2-4-8-13(11)14/h1-8H,9H2

InChI key

RJGDLRCDCYRQOQ-UHFFFAOYSA-N

Application

  • Surfactant modified electrochemical sensor for determination of Anthrone–A cyclic voltammetry: Research on developing an electrochemical sensor modified by surfactants for the determination of Anthrone, emphasizing its potential in environmental monitoring (NS Prinith, JG Manjunatha, 2019).

Analysis Note

Assay (LC-MS): ≥ 97.0 % (a/a)
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service