Skip to Content
Merck
All Photos(3)

Key Documents

V900920

Sigma-Aldrich

L-(+)-Arabinose

Vetec, reagent grade, ≥99%

Synonym(s):

Aldehydo-L-arabinose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
Beilstein:
1723085
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

≥99%

mp

160-163 °C (lit.)

storage temp.

room temp

SMILES string

OC[C@H](O)[C@H](O)[C@@H](O)C=O

InChI

1S/C5H10O5/c6-1-3(8)5(10)4(9)2-7/h1,3-5,7-10H,2H2/t3-,4-,5+/m0/s1

InChI key

PYMYPHUHKUWMLA-VAYJURFESA-N

Application


  • Halopseudomonas species: Cultivation and molecular genetic tools.: This study describes the application of L-(+)-Arabinose in the cultivation of Halopseudomonas species, highlighting its role in microbial biotechnology and genetic tool development (Kruse et al., 2024).

  • easyPACId, a Simple Method for Induced Production, Isolation, Identification, and Testing of Natural Products from Proteobacteria.: Discusses a methodology that includes the use of L-(+)-Arabinose for inducing the production of natural products in Proteobacteria, demonstrating its value in microbial natural product research (Bode et al., 2023).

Biochem/physiol Actions

L-Arabinose is the naturally occurring isomer and is a constituent of plant polysaccharides. Most bacteria contain an inducible arabinose operon that codes for a series of enzymes and transporters that allows L-arabinose to be used as the sole carbon source in microbial culture.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiaorong Tan et al.
Journal of diabetes research, 2019, 5483946-5483946 (2019-06-22)
This study is aimed at evaluating the effects, functions, and mechanism of HNF1α on hepatic glycolipid metabolism. In this study, free fatty acid- (FFA-) induced steatosis of hepatocyte liver cell LO2 was used as an in vitro model. The methods
Naeem Anwar et al.
PloS one, 9(8), e106095-e106095 (2014-08-26)
In Salmonella enterica serovar Typhimurium (S. Typhimurium), biofilm-formation is controlled by the cytoplasmic intracellular small-molecular second messenger cyclic 3', 5'-di- guanosine monophosphate (c-di-GMP) through the activities of GGDEF and EAL domain proteins. Here we describe that deleting either dsbA or
Wei Sun et al.
Infection and immunity, 82(10), 4390-4404 (2014-08-13)
Here, we constructed a Yersinia pseudotuberculosis mutant strain with arabinose-dependent regulated and delayed shutoff of crp expression (araC P(BAD) crp) and replacement of the msbB gene with the Escherichia coli msbB gene to attenuate it. Then, we inserted the asd
Hua Tian et al.
International journal of biological macromolecules, 79, 577-582 (2015-06-01)
Two polysaccharides (ULP1 and ULP2) were isolated through ultrasonic-assisted extraction from green seaweed Ulva lactuca L. which was collected from the South China Sea. The highest yield of 17.57% was obtained under the conditions of 2% NaOH, 90 °C, material/water
Nicole Acosta et al.
Journal of bacteriology, 197(2), 262-276 (2014-11-05)
The Cpx pathway, a two-component system that employs the sensor histidine kinase CpxA and the response regulator CpxR, regulates crucial envelope stress responses across bacterial species and affects antibiotic resistance. To characterize the CpxR regulon in Vibrio cholerae, the transcriptional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service