Skip to Content
Merck
All Photos(1)

Key Documents

V900847

Supelco

Acrylamide/Bis-acrylamide

Vetec, reagent grade, 19:1, powder, suitable for electrophoresis

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

product line

Vetec

form

powder

concentration

19:1

suitability

suitable for electrophoresis

SMILES string

NC(=O)C=C.C=CC(=O)NCNC(=O)C=C

InChI

1S/C7H10N2O2.C3H5NO/c1-3-6(10)8-5-9-7(11)4-2;1-2-3(4)5/h3-4H,1-2,5H2,(H,8,10)(H,9,11);2H,1H2,(H2,4,5)

InChI key

OCQZXMCGTAWGEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ready to reconstitute dry powder blends are accurately pre-blended to produce a 40% (w/v) stock solution for use in protein and nucleic acid electrophoresis. The concentration is based on the total weight of both the acrylamide and bis-acrylamide.

Features and Benefits

Eliminates the need to weigh toxic acrylamide and bis-acrylamide.

Reconstitution

Reconstitute by slowly adding the indicated amount of distilled water to the container and dissolve contents.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service