Skip to Content
Merck
All Photos(1)

Key Documents

V900843

Sigma-Aldrich

Tris-Borate-EDTA buffer

Vetec, reagent grade, 10 ×, solution, RNase and DNase free

Synonym(s):

TBE buffer

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41105319
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

form

solution

feature

RNase and DNase free

concentration

10 ×

SMILES string

OB(O)O.NC(CO)(CO)CO.OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C10H16N2O8.C4H11NO3.BH3O3/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;5-4(1-6,2-7)3-8;2-1(3)4/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);6-8H,1-3,5H2;2-4H

InChI key

OSBLTNPMIGYQGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

TBE running buffer is the most commonly used buffer for DNA and RNA polyacrylamide gel electrophoresis. TBE is used with non-denaturing or denaturing (7 M urea) gels. It is also routinely used for DNA automated sequencing gel. TBE can also be used for agarose gels but is not recommended for preparative gels for recovery of nucleic acids.
Dilution of the TBE stock concentrates to a 1× TBE running buffer results in a buffer containing 89 mM Tris-borate and 2 mM EDTA, pH 8.3. The 5× or 10× stocks may also be added to an acrylamide/bis-acrylamide stock solution for making the PAGE gel. Applied voltages of less than 5 V/cm (distance between the electrodes of the unit) are recommended for maximum resolution.

Preparation Note

Prepared with Biotechnology Performance Certified Trizma base (Product Code T6066) and Molecular Biology Reagents boric acid (Product Code B6768) and EDTA disodium salt (Product Code E5134).

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service