Skip to Content
Merck
All Photos(3)

Key Documents

V900767

Sigma-Aldrich

2,4-Dihydroxybenzoic acid

Vetec, reagent grade, 97%

Synonym(s):

β-Resorcylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CO2H
CAS Number:
Molecular Weight:
154.12
Beilstein:
1946213
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

97%

mp

208-211 °C (dec.) (lit.)

SMILES string

OC(=O)c1ccc(O)cc1O

InChI

1S/C7H6O4/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3,8-9H,(H,10,11)

InChI key

UIAFKZKHHVMJGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R F Anderson et al.
The Journal of biological chemistry, 262(36), 17475-17479 (1987-12-25)
The spectra of radicals formed upon the addition of .OH radicals to the 3 position of substrates for p-hydroxybenzoate hydroxylase (4-hydroxybenzoic acid, 2,4-dihydroxybenzoic acid, and 4-aminobenzoic acid) have been determined using pulse radiolysis combined with the results from high performance
R F Anderson et al.
The Journal of biological chemistry, 266(20), 13086-13090 (1991-07-15)
Combined optical and conductimetric measurements in aqueous solution indicate that at high pH (greater than or equal to 10).OH radicals react with the phenoxide form of 2,4-dihydroxybenzoic acid to form transiently phenoxyl radicals and a small amount of hydroxyeyclohexadienyl (HCHD)
Liangxiong Xu et al.
Journal of natural products, 73(5), 885-889 (2010-05-01)
Six new beta-resorcylic acid lactones (1-6), named paecilomycins A-F, and five known compounds, aigilomycin B (7), zeaenol (8), aigialomycin D (9), aigialomycin F (10), and aigialospirol, were isolated from the mycelial solid culture of Paecilomyces sp. SC0924. Their structures were
Anup Kollanoor Johny et al.
Foodborne pathogens and disease, 7(10), 1165-1170 (2010-07-14)
This study investigated the efficacy of plant-derived antimicrobials, namely, trans-cinnamaldehyde, β-resorcylic acid, carvacrol, thymol, and eugenol or their combination, in increasing the sensitivity of Salmonella Typhimurium DT104 to five antibiotics. The subinhibitory concentrations of each antimicrobial or their combination containing
Kamal Azrague et al.
Journal of hazardous materials, 237-238, 71-78 (2012-09-15)
2,4-Dihydroxybenzoic acid (2,4-DHBA) is found frequently as a pollutant in natural waters and represents a threat to water quality because it is a precursor to the formation of quinones which are highly toxic. The degradation of 2,4-DHBA using the vacuum

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service