V900740
4-Aminosalicylic acid
99%, Vetec™
Synonym(s):
4-Amino-2-hydroxybenzoic acid, PAS
About This Item
Product Name
4-Aminosalicylic acid, Vetec™, reagent grade, 99%
grade
reagent grade
product line
Vetec™
Assay
99%
form
powder
color
white to beige
mp
135-145 °C (lit.)
SMILES string
Nc1ccc(C(O)=O)c(O)c1
InChI
1S/C7H7NO3/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3,9H,8H2,(H,10,11)
InChI key
WUBBRNOQWQTFEX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service